Learn More
Mac-Mod Avantor Genesis AQ, 120 Å, 4 µm, 4.6 x 100 mm HPLC Column
SDP
Catalog No. 5000143589
Encompass
Avantor Genesis phases are based on high purity metal-free spherical silica.
Catalog No. 50-001-43589 Supplier Mac-Mod Supplier No. FM10951E
Avantor Genesis phases are based on high purity metal-free spherical silica.