Learn More
Mac-Mod Avantor Genesis C18, 120 Å, 3 µm, 4.6 x 150 mm HPLC Column
SDP
Catalog No. 5000143769
Encompass
Avantor Genesis phases are based on high purity metal-free spherical silica.
Catalog No. 50-001-43769 Supplier Mac-Mod Supplier No. FM15963E / 5140908
Avantor Genesis phases are based on high purity metal-free spherical silica.