Learn More
Medchemexpress LLC HPMC (Viscosity: 11250-21000 mPa.s) | 9004-65-3 | 500 MG
SDP
Catalog No. 5000433546
Encompass_Preferred
HPMC (Viscosity:11250-21000mPa.s) is a hydrophilic, non-ionic cellulose ether used to form swellable-soluble matrices. It provides controlled release of a drug, effectively increasing the duration of release to prolong its therapeutic effect. When used in matrix tablets, it hydrates on contact with water to produce a viscous gel barrier.
  • Hydrophilic, non-ionic cellulose ether
  • Forms swellable-soluble matrices
  • Provides controlled release of drugs
  • Increases duration of drug release
  • Forms a viscous gel barrier
  • Effective in improving blood glucose metabolism
  • Suppresses oxidative stress
Catalog No. 50-004-33546 Supplier Medchemexpress LLC Supplier No. HYA0104500MG
HPMC (Viscosity:11250-21000mPa.s) is a hydrophilic, non-ionic cellulose ether used to form swellable-soluble matrices. It provides controlled release of a drug, effectively increasing the duration of release to prolong its therapeutic effect. When used in matrix tablets, it hydrates on contact with water to produce a viscous gel barrier.
  • Hydrophilic, non-ionic cellulose ether
  • Forms swellable-soluble matrices
  • Provides controlled release of drugs
  • Increases duration of drug release
  • Forms a viscous gel barrier
  • Effective in improving blood glucose metabolism
  • Suppresses oxidative stress