Learn More
Waters Corp Waters Tool Kit, HPMV, Seal Installation
Catalog No. 50466773 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides the specialized tools required for installing seals within HP Valve Rebuild Kits. It is essential for ensuring the correct installation of high-pressure motorized valves, helping to prevent potential repair costs from improper assembly.
  • Includes a seal punch, seal guide, and wood screw.
  • Ensures proper installation of high-pressure motorized valves.
  • Helps prevent incorrect installation which can lead to greater repair costs.
  • Designed for use with high pressure motorized valve rebuild kits.
Catalog No. 50-466-773 Supplier Waters Corp Supplier No. WAT045427
This kit provides the specialized tools required for installing seals within HP Valve Rebuild Kits. It is essential for ensuring the correct installation of high-pressure motorized valves, helping to prevent potential repair costs from improper assembly.
  • Includes a seal punch, seal guide, and wood screw.
  • Ensures proper installation of high-pressure motorized valves.
  • Helps prevent incorrect installation which can lead to greater repair costs.
  • Designed for use with high pressure motorized valve rebuild kits.