Learn More
TARGETMOL CHEMICALS INC HPOB 5MG
Supplier: TARGETMOL CHEMICALS INC T24305MG
Also available in 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HPOB is an effective and specific HDAC6 inhibitor (IC50 56 nM) >30-fold selectivity over other HDACs. purity: 99%