Learn More
Medchemexpress LLC HY-19747 5mg Medchemexpress, HPOB CAS:1429651-50-2 Purity:>98%
SDP
Catalog No. 502082172
Encompass_Preferred
Medchemexpress, HY-19747 5mg HPOB CAS:1429651-50-2 HPOB is a highly potent and selective inhibitor of HDAC6 with an IC50 of 56 nM. HPOB displays >30 fold less potent against other HDACs. HPOB enhances the effectiveness of DNA-damaging anticancer agents in transformed cells but not normal cells. HPOB does not block the ubiquitin-binding activity of HDAC6. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2172 Supplier Medchemexpress LLC Supplier No. HY197475MG
Medchemexpress, HY-19747 5mg HPOB CAS:1429651-50-2 HPOB is a highly potent and selective inhibitor of HDAC6 with an IC50 of 56 nM. HPOB displays >30 fold less potent against other HDACs. HPOB enhances the effectiveness of DNA-damaging anticancer agents in transformed cells but not normal cells. HPOB does not block the ubiquitin-binding activity of HDAC6. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.