Learn More
MSC PRO-SAFE G-Tek PolyKor Xrystal Cut-Resistant Gloves, Dyneema, A4, X-Small, 1 Pair
Catalog No. 5000687364
Encompass
These cut-resistant gloves provide hand protection in demanding environments, featuring a durable construction with an ANSI/ISEA A4 cut resistance level. The design incorporates a palm and finger coating for enhanced grip and tactile sensitivity, making them suitable for tasks requiring high dexterity.
  • ANSI/ISEA A4 cut resistance
  • Dyneema and HPPE material construction
  • Seamless lining for comfort
  • NeoFoam palm and finger coating
  • Knit cuff style
  • Resistant to chemicals, water, and ultraviolet light
  • Silicone-free and washable design
Catalog No. 50-006-87364 Supplier MSC Supplier No. 33359001
These cut-resistant gloves provide hand protection in demanding environments, featuring a durable construction with an ANSI/ISEA A4 cut resistance level. The design incorporates a palm and finger coating for enhanced grip and tactile sensitivity, making them suitable for tasks requiring high dexterity.
  • ANSI/ISEA A4 cut resistance
  • Dyneema and HPPE material construction
  • Seamless lining for comfort
  • NeoFoam palm and finger coating
  • Knit cuff style
  • Resistant to chemicals, water, and ultraviolet light
  • Silicone-free and washable design