Learn More
MSC Honeywell Perfect Fit 3.0 Gloves, Cut-Resistant, 18-Gauge, Antistatic, 10/Pk.
Catalog No. 5000510021
Encompass
These 18-gauge, cut-resistant work gloves are engineered to provide workers with enhanced comfort, dexterity, and protection. Featuring a seamless HPPE, nylon, carbon, and elastane liner with a breathable polyurethane palm coating, these gloves are designed for high-performance handling in various industrial applications while maintaining a lightweight feel.
  • Glass fiber-free and silicone-free design
  • Antistatic properties meeting EN 16350 standards
  • Touchscreen capable material for operational efficiency
  • Certified for safe food handling
  • Low DMF content of less than 100 ppm
  • OEKO-TEX® 100 certified
  • Washable up to 3 washing cycles
  • ANSI/ISEA cut resistance level A2 and abrasion resistance level 3
  • Clear markings for size and batch identification
Catalog No. 50-005-10021 Supplier MSC Supplier No. 05764774
These 18-gauge, cut-resistant work gloves are engineered to provide workers with enhanced comfort, dexterity, and protection. Featuring a seamless HPPE, nylon, carbon, and elastane liner with a breathable polyurethane palm coating, these gloves are designed for high-performance handling in various industrial applications while maintaining a lightweight feel.
  • Glass fiber-free and silicone-free design
  • Antistatic properties meeting EN 16350 standards
  • Touchscreen capable material for operational efficiency
  • Certified for safe food handling
  • Low DMF content of less than 100 ppm
  • OEKO-TEX® 100 certified
  • Washable up to 3 washing cycles
  • ANSI/ISEA cut resistance level A2 and abrasion resistance level 3
  • Clear markings for size and batch identification