Learn More
MSC PIP Cut, Puncture, And Abrasion-Resistant Gloves, 18 ga, Cut A4, Size X-Large, 1 Pair
Catalog No. 5000510027
Encompass
These cut, puncture, and abrasion-resistant gloves feature a lightweight 18-gauge seamless liner engineered for tactile sensitivity and dexterity. Constructed from a blend of HPPE, nylon, carbon, and elastane, these gloves offer reliable protection with a breathable polyurethane palm coating that enhances grip. Designed for precision handling, they are antistatic, touchscreen-capable, and silicone-free, making them suitable for diverse industrial environments.
  • ANSI/ISEA cut resistance level A4
  • ANSI/ISEA puncture resistance level 3
  • ANSI/ISEA abrasion resistance level 3
  • Breathable polyurethane palm coating
  • Antistatic and touchscreen capable
  • Certified for food handling per FDA 21CFR177.2600
  • Silicone-free and glass fiber-free construction
  • Color-coded markings for size identification
  • Washable for extended service life
  • Low DMF content (<100 ppm)
Catalog No. 50-005-10027 Supplier MSC Supplier No. 05770334
These cut, puncture, and abrasion-resistant gloves feature a lightweight 18-gauge seamless liner engineered for tactile sensitivity and dexterity. Constructed from a blend of HPPE, nylon, carbon, and elastane, these gloves offer reliable protection with a breathable polyurethane palm coating that enhances grip. Designed for precision handling, they are antistatic, touchscreen-capable, and silicone-free, making them suitable for diverse industrial environments.
  • ANSI/ISEA cut resistance level A4
  • ANSI/ISEA puncture resistance level 3
  • ANSI/ISEA abrasion resistance level 3
  • Breathable polyurethane palm coating
  • Antistatic and touchscreen capable
  • Certified for food handling per FDA 21CFR177.2600
  • Silicone-free and glass fiber-free construction
  • Color-coded markings for size identification
  • Washable for extended service life
  • Low DMF content (<100 ppm)