Learn More
MSC PIP WorkEasy Cut-Resistant Gloves, A3, 13 ga, 10/Pk.
Catalog No. 5000509164
Encompass
These cut, puncture and abrasion-resistant gloves provide reliable hand protection in mechanical environments. Designed with a seamless HPPE and fiberglass liner, they offer a 13 gauge construction for a balance of dexterity and durability. The palm and fingertips feature a smooth polyurethane coating, ensuring a secure grip while maintaining breathability.
  • ANSI/ISEA cut resistance level A3 for protection against sharp objects
  • ANSI/ISEA abrasion resistance level 4 for long-lasting wear
  • ANSI/ISEA puncture resistance level 2
  • Seamless knit lining for enhanced comfort
  • Extended cuff design for additional wrist protection
  • Polyurethane palm coating provides reliable grip in varied conditions
Catalog No. 50-005-09164 Supplier MSC Supplier No. 97380760
These cut, puncture and abrasion-resistant gloves provide reliable hand protection in mechanical environments. Designed with a seamless HPPE and fiberglass liner, they offer a 13 gauge construction for a balance of dexterity and durability. The palm and fingertips feature a smooth polyurethane coating, ensuring a secure grip while maintaining breathability.
  • ANSI/ISEA cut resistance level A3 for protection against sharp objects
  • ANSI/ISEA abrasion resistance level 4 for long-lasting wear
  • ANSI/ISEA puncture resistance level 2
  • Seamless knit lining for enhanced comfort
  • Extended cuff design for additional wrist protection
  • Polyurethane palm coating provides reliable grip in varied conditions