Learn More
MSC PIP Cut-Resistant Gloves, ANSI A5, 18 Gauge, Small, 1 Pair
Catalog No. 5000509431
Encompass
These cut, puncture, and abrasion-resistant gloves feature an 18-gauge seamless liner engineered from a blend of carbon, elastane, HPPE, steel, and polyester, finished with a black polyurethane palm coating. Designed for high-performance safety, these gloves combine advanced dexterity with robust protection in various industrial applications.
  • ANSI/ISEA cut resistance level A5
  • ANSI/ISEA abrasion resistance level 3
  • ANSI/ISEA puncture resistance level 3
  • Polyurethane coated palm for high grip performance
  • Touchscreen capable
  • Antistatic compliant per EN 16350
  • Certified for food handling per FDA 21CFR177.2600 and EC 1935/2004
  • OEKO-TEX 100 certified, silicone-free, and glass-fiber-free
  • Low DMF content (<100 ppm)
  • Washable up to 3 cycles
Catalog No. 50-005-09431 Supplier MSC Supplier No. 41901737
These cut, puncture, and abrasion-resistant gloves feature an 18-gauge seamless liner engineered from a blend of carbon, elastane, HPPE, steel, and polyester, finished with a black polyurethane palm coating. Designed for high-performance safety, these gloves combine advanced dexterity with robust protection in various industrial applications.
  • ANSI/ISEA cut resistance level A5
  • ANSI/ISEA abrasion resistance level 3
  • ANSI/ISEA puncture resistance level 3
  • Polyurethane coated palm for high grip performance
  • Touchscreen capable
  • Antistatic compliant per EN 16350
  • Certified for food handling per FDA 21CFR177.2600 and EC 1935/2004
  • OEKO-TEX 100 certified, silicone-free, and glass-fiber-free
  • Low DMF content (<100 ppm)
  • Washable up to 3 cycles