Learn More
MSC SHOWA Cut and Abrasion-Resistant Gloves, HPPE, 18 ga, X-Large, 1/Pr.
Catalog No. 5000684281
Encompass
These cut and abrasion-resistant gloves provide superior hand protection in demanding environments. Constructed from an 18-gauge seamless knit HPPE liner, they offer a balance of cut resistance and dexterity for precise handling tasks.
  • Features a polyurethane palm and fingertip coating for enhanced grip and protection
  • Offers ANSI/ISEA cut resistance level A4 and abrasion resistance level 4
  • Embossed grip surface improves control in various conditions
  • Lightweight, breathable, and form-fitting design ensures comfort during extended use
  • Certified to OEKO-TEX® STANDARD 100
  • Includes elastic cuffs for a secure, ergonomic fit
Catalog No. 50-006-84281 Supplier MSC Supplier No. 61934295
These cut and abrasion-resistant gloves provide superior hand protection in demanding environments. Constructed from an 18-gauge seamless knit HPPE liner, they offer a balance of cut resistance and dexterity for precise handling tasks.
  • Features a polyurethane palm and fingertip coating for enhanced grip and protection
  • Offers ANSI/ISEA cut resistance level A4 and abrasion resistance level 4
  • Embossed grip surface improves control in various conditions
  • Lightweight, breathable, and form-fitting design ensures comfort during extended use
  • Certified to OEKO-TEX® STANDARD 100
  • Includes elastic cuffs for a secure, ergonomic fit