Learn More
Medchemexpress LLC HPRG Human HEK293 10ug | 10ug
SDP
Catalog No. 5000186802
Encompass_Preferred
HPRG protein is a multifunctional plasma glycoprotein with the ability to bind multiple ligands including heme heparin thrombospondin etc HPRG regulates immune processes angiogenesis coagulation and fibrinolysis through zinc-dependent binding to heparin and glycosaminoglycans HPRG Protein Human (HEK293 His) is the recombinant human-derived HPRG protein expressed by HEK293 with C-His C-10 His labeled tag
Catalog No. 50-001-86802 Supplier Medchemexpress LLC Supplier No. HYP7738010UG
HPRG protein is a multifunctional plasma glycoprotein with the ability to bind multiple ligands including heme heparin thrombospondin etc HPRG regulates immune processes angiogenesis coagulation and fibrinolysis through zinc-dependent binding to heparin and glycosaminoglycans HPRG Protein Human (HEK293 His) is the recombinant human-derived HPRG protein expressed by HEK293 with C-His C-10 His labeled tag