Learn More
Medchemexpress LLC HPRG Human HEK293 5ug | 5ug
SDP
Catalog No. 5000187258
Encompass_Preferred
HPRG protein is a multifunctional plasma glycoprotein with the ability to bind multiple ligands including heme heparin thrombospondin etc HPRG regulates immune processes angiogenesis coagulation and fibrinolysis through zinc-dependent binding to heparin and glycosaminoglycans HPRG Protein Human (HEK293 His) is the recombinant human-derived HPRG protein expressed by HEK293 with C-His C-10 His labeled tag
Catalog No. 50-001-87258 Supplier Medchemexpress LLC Supplier No. HYP773805UG
HPRG protein is a multifunctional plasma glycoprotein with the ability to bind multiple ligands including heme heparin thrombospondin etc HPRG regulates immune processes angiogenesis coagulation and fibrinolysis through zinc-dependent binding to heparin and glycosaminoglycans HPRG Protein Human (HEK293 His) is the recombinant human-derived HPRG protein expressed by HEK293 with C-His C-10 His labeled tag