Learn More
Aviva Systems Biology Corp Hprt Antibody - C-terminal region (ARP61048_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61048_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hprt. It was validated on Western Blot. . Alternate names: C81579, HPGRT, Hprt1, MGC103149ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-239