Learn More
Aviva Systems Biology Corp HPRT1 Antibody - middle region (ARP48471_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48471_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HPRT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HPRT1. Alternate names: HGPRT, HPRTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59835