Learn More
MyBioSource Rat Hypoxanthine Guanine Phosphoribosyltransferase ELISA Kit
SDP
Supplier:  MyBioSource MBS201947796STRIPWELLS
Encompass_Preferred
The MyBioSource Rat Hypoxanthine Guanine Phosphoribosyltransferase ELISA Kit is a sandwich enzyme immunoassay for the in vitro quantitative measurement of HPRT in rat serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. The kit has a detection range of 0.156 ng/mL - 10 ng/mL. It contains a pre-coated 96-well strip plate, standard, biotinylated detection Ab, HRP conjugate, TMB substrate, stop solution and other reagents. The kit has high sensitivity and excellent specificity for detection of rat HPRT.
Catalog No. NC2586410