Learn More
MyBioSource HPRT1 ELISA Kit
SDP
Supplier:  MyBioSource MBS720149996STRIPWELLS
Encompass_Preferred
The MyBioSource HPRT1 ELISA Kit quantifies human HPRT1 in various biological fluids using a 96-well plate coated with a specific antibody. Samples are added, followed by a biotinylated detection antibody, HRP-conjugated streptavidin, and TMB substrate, resulting in a color change measured at 450 nm.
Catalog No. NC2586406