Learn More
SPEX CERTIPREP LLC 3P HPS INORGANIC 100ML START KIT
Catalog No. NC3454951
Encompass
Starter Kit D: Contains 3 x 100mL single element bottles at 1000u g/mL each in 2-5%HCl. Au, Pd, Pt included.
Catalog No. NC3454951 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICPKITD100
Starter Kit D: Contains 3 x 100mL single element bottles at 1000u g/mL each in 2-5%HCl. Au, Pd, Pt included.