Learn More
SPEX CERTIPREP LLC 3P HPS INORGANIC 250ML START KIT
Catalog No. NC3454954
Encompass
STARTER KIT D: INCLUDES 3 x 250ML SINGLE ELEMENT BOTTLES AT 1000 UG/ML EA IN 2-5% HCl. Au, Pd, Pt.
Catalog No. NC3454954 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICPKITD250
STARTER KIT D: INCLUDES 3 x 250ML SINGLE ELEMENT BOTTLES AT 1000 UG/ML EA IN 2-5% HCl. Au, Pd, Pt.