Learn More
Aviva Systems Biology Corp HPSE Antibody - N-terminal region (ARP60765_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60765_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HPSE. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human HPSE. Alternate names: HPA, HPR1, HPSE1, HSE1, HPA1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-153