Learn More
AdipoGen HPTS
SDP
Catalog No. 502560787
Encompass_Preferred
Chemical. CAS 6358-69-627928-00-3 free acid. Formula C16H7Na3O10S3. MW 524.39. Synthetic. Useful for optical sensor applications. Fluorescent pH indicator for physiological range.
Catalog No. 50-256-0787 Supplier AdipoGen Supplier No. CDXH0034G001
Chemical. CAS 6358-69-627928-00-3 free acid. Formula C16H7Na3O10S3. MW 524.39. Synthetic. Useful for optical sensor applications. Fluorescent pH indicator for physiological range.