Learn More
Creative Biomart Recombinant HPV-16 Antigen, GST-tagged
Supplier:  Creative Biomart HPV345
Encompass_Preferred
The Recombinant HPV-16 antigen is a full length protein expressed in E. coli having an Mw of 56.7 kDa. The protein is fused to a 26 kDa GST-Tag, having a total Mw of 78 kDa and purified by standard ch
Catalog No. 50-208-4723