Learn More
Abnova™ HPV Screening CISH Probe
Catalog No. 89335577 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
Catalog No. For Use With (Application)
89-335-577 CISH-P
1 options
Catalog No. 89-335-577 Supplier Abnova™ Supplier No. CG0026
Add to Cart
Add to Cart
HPV Screening CISH Probe is designed for the qualitative detection of human papillomavirus (HPV) type 6/11/16/18/31/33/35/39/45/51/52/56/58/59/66/68/82 DNA in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
The probe is intended to be used in combination with the CISH Implementation HRP-DAB Kit, which provides necessary reagents for specimen pretreatment and post-hybridization processing.
The staining pattern in the nucleus can be observed as discrete and dot-shaped signals in case of integrated HPV, or as a strong and homogeneous nuclear staining in case of episomal HPV. A cytoplasmic staining is observed when RNA sequences of HPV are detected. Visualization of signals should be performed using a set of objectives ranging from an at least 200-fold to 630-fold magnification. The presence of the episomal staining pattern is usually detected clearly by an objective with 200-fold magnification, whereas the detection of the integrated HPV pattern requires a greater magnification, preferably 630-fold. Do not evaluate areas of necrosis, overlapping nuclei, over-digested nuclei and nuclei with weak signal intensity.
Specifications
Content And Storage The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (37°C) and mix briefly before use.
For Use With (Application) CISH-P