Learn More
Enzo Life Sciences PATHO-GENE HPV type 16 18 probe (6ml)
SDP
Supplier:  Enzo Life Sciences ENZ328866000
Encompass_Preferred
The PATHO-GENE HPV type 16/18 probe is a mixture of biotin-labeled HPV 16 and HPV 18-specific probes in buffered formamide and hybridization enhancers. Long Term Storage: +4°C.
Catalog No. 50-201-2496