Learn More
Enzo Life Sciences PATHO-GENE HPV type 6 11 probe (1ml)
SDP
Supplier:  Enzo Life Sciences ENZ32885
Encompass_Preferred
The PATHO-GENE HPV type 6/11 probe is a mixture of biotin-labeled HPV 6 and HPV 11-specific probes in buffered formamide and hybridization enhancers. Long Term Storage: +4°C.
Catalog No. 50-201-2494