Learn More
Diagnostic Biosystems HPV16 HPV 16
Catalog No. 502407636
Encompass_Preferred
This antibody stains with HPV type 16. L1 protein is a major capsid protein of HPV type 16. HPV 6 and 11 have been associated with relatively benign diseases such as genital warts but type 16 and 18 are strongly associated with cervical, vaginal and vulvar malignancies.
Catalog No. 50-240-7636 Supplier Diagnostic Biosystems Supplier No. PDM166
This antibody stains with HPV type 16. L1 protein is a major capsid protein of HPV type 16. HPV 6 and 11 have been associated with relatively benign diseases such as genital warts but type 16 and 18 are strongly associated with cervical, vaginal and vulvar malignancies.