Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM1502XXP1
Encompass_Preferred
Product Type: Antibody. Application: IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM405. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: HPV-16 infected cells or cervical tissue. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant major capsid protein L1 from human papilloma virus type 16. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5417