Learn More
Biotium HPV16 E1/E4 (Human Papilloma Virus 16)(HPV16 E1/E4), CF568 conjugate, 0.1mg/mL
Supplier:  Biotium BNC683066500
Encompass_Preferred
The human papilloma virus (HPV) family of DNA tumor viruses includes HPV16, a 'high-risk' sexually-transmitted HPV that can lead to cervical, anal, vulvar, head, neck, and penile cancer. The HPV16E1/E4 protein is expressed abundantly in cells supporting viral DNA amplification, but is lost during malignant progression. HPV16E1/E4 causes G2 cell cycle arrest by associating with and preventing the nuclear entry of Cdk1/cyclin B1 complexes. HPV16E1/E4 also interacts with cyclin A and Cdk2 during the G2 phase of the cell cycle, and this association may increase the efficiency with which HPV16E1/E4 is able to prevent mitotic entry. HPV16E1/E4 also associates with keratin intermediate filaments and causes the network to collapse. Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for d
Catalog No. 50-006-17103