Learn More
Biotium HPV16 E2 (Human Papilloma Virus 16)(TVG 261), CF488A conjugate, 0.1mg/mL
Supplier:  Biotium BNC883073500
Encompass_Preferred
The human papilloma virus (HPV) family of DNA tumor viruses includes HPV16, a strain that is responsible for the largest number of cases of cervical cancers linked to the family. HPV16E1 and HPV16E2 are proteins that are involved in the regulation of viral DNA replication and are important for infected cell homeostasis. HPV16E2 specifically regulates the expression of the E6 and E7 oncoproteins by binding to four sites within the viral long control region, possibly involving interactions with nuclear hormone receptors. Integration of the HPV genome into the host DNA usually disrupts the HPV16E2 gene open reading frames, resulting in an overexpression of E6 and E7 genes, an event that may lead to the malignant transformation of cervical cancer. HPV16E2 is also able to induce apoptotic cell death via two pathways: the first through the binding of p53 and the second through the binding of the viral genome. Primary antibodies are available purified, or with a selection of fluores
Catalog No. 50-006-19651