Learn More
Gentarget HPV16 E6-E7 EXPRESSION LENTIVI
Catalog No. NC3585485
Encompass
Premade Lentivirus expresses HPV16 E6-E7 fusion viral antigen /click to see its coding sequence/ under enhanced CMV promoter. Its coding sequence match to CD regions of E6 and E7 in human papillomavirus type 16 /HPV16/ Accession ID/ LC456637/.The lentivirus contains RFP Blasticidin dual selection which allows to select the transduced cells via Blasticidin antibiotic or via cell sorting by RFP fluorescent signal.
Catalog No. NC3585485 Supplier Gentarget Supplier No. LVP1488PBS
Premade Lentivirus expresses HPV16 E6-E7 fusion viral antigen /click to see its coding sequence/ under enhanced CMV promoter. Its coding sequence match to CD regions of E6 and E7 in human papillomavirus type 16 /HPV16/ Accession ID/ LC456637/.The lentivirus contains RFP Blasticidin dual selection which allows to select the transduced cells via Blasticidin antibiotic or via cell sorting by RFP fluorescent signal.