Learn More
Sino Biological HPV16 E6 Protein (His Tag)
Supplier:  Sino Biological 40963V07E100
Encompass_Preferred
A DNA sequence encoding the HPV16 E6 (P03126) (Met1-Leu158) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3571