Learn More
Sino Biological HPV18 E6 Protein (N-His-MBP & C-AVI Tag)
Supplier:  Sino Biological 40964V79E100
Encompass_Preferred
A DNA sequence encoding the HPV18 E6 (P06463) (Met1-Val158) was fused with an N-terminal polyhistidine-tagged MBP tag at the N-terminus and an AVI tag at the C-terminus.
Catalog No. 50-255-3570