Learn More
ATCC HPV-52; Molecular Clone; Plasmid: pCD15
Catalog No. 502382635
Encompass
This is a full length clone of the human papilloma virus 52 (HPV-52), linearized at the EcoRI site and cloned into the EcoRI site of the pUC19 vector.
Catalog No. 50-238-2635 Supplier ATCC Supplier No. VRMC29
This is a full length clone of the human papilloma virus 52 (HPV-52), linearized at the EcoRI site and cloned into the EcoRI site of the pUC19 vector.