Learn More
Bio Rad Laboratories HPX-87H COLUMN
Catalog No. NC9265562
Encompass
Activated affinity media for spontaneous ligand immobilization allow creation of an affinity matrix of choice by coupling a ligand such as an antibody, enzyme, antigen, or receptor. Affi-Gel 10 support couples ligands via primary amines.
Catalog No. NC9265562 Supplier Bio Rad Laboratories Supplier No. 1250140
Activated affinity media for spontaneous ligand immobilization allow creation of an affinity matrix of choice by coupling a ligand such as an antibody, enzyme, antigen, or receptor. Affi-Gel 10 support couples ligands via primary amines.