Learn More
New England Biolabs, Inc. Hpy166II – 1000 units
SDP
Catalog No. 50995256
Encompass
A restriction endonuclease that recognizes the sequence GTN^_NAC.
Catalog No. 50-995-256 Supplier New England Biolabs, Inc. Supplier No. R0616S
A restriction endonuclease that recognizes the sequence GTN^_NAC.