Learn More
New England Biolabs, Inc. Hpy188I – 1000 units
SDP
Catalog No. 50812354
Encompass
A restriction endonuclease that recognizes the sequence TC_N^GA.
Catalog No. 50-812-354 Supplier New England Biolabs, Inc. Supplier No. R0617S
A restriction endonuclease that recognizes the sequence TC_N^GA.