Learn More
New England Biolabs, Inc. Hpy188I – 5000 units
SDP
Catalog No. 50812353
Encompass
A restriction endonuclease that recognizes the sequence TC_N^GA.
Catalog No. 50-812-353 Supplier New England Biolabs, Inc. Supplier No. R0617L
A restriction endonuclease that recognizes the sequence TC_N^GA.