Learn More
New England Biolabs, Inc. Hpy188III – 500 units
SDP
Catalog No. 50812362
Encompass
A restriction endonuclease that recognizes the sequence TC^NN_GA.
Catalog No. 50-812-362 Supplier New England Biolabs, Inc. Supplier No. R0622S
A restriction endonuclease that recognizes the sequence TC^NN_GA.