Learn More
New England Biolabs, Inc. HpyAV – 100 units
SDP
Catalog No. 50995258
Encompass
A restriction endonuclease that recognizes the sequence CCTTCNNNNN_N^.
Catalog No. 50-995-258 Supplier New England Biolabs, Inc. Supplier No. R0621S
A restriction endonuclease that recognizes the sequence CCTTCNNNNN_N^.