Learn More
New England Biolabs, Inc. HpyCH4III – 250 units
SDP
Catalog No. 50812356
Encompass
A restriction endonuclease that recognizes the sequence AC_N^GT.
Catalog No. 50-812-356 Supplier New England Biolabs, Inc. Supplier No. R0618S
A restriction endonuclease that recognizes the sequence AC_N^GT.