Learn More
New England Biolabs, Inc. HpyCH4IV – 2500 units
SDP
Catalog No. 50812357
Encompass
A restriction endonuclease that recognizes the sequence A^CG_T.
Catalog No. 50-812-357 Supplier New England Biolabs, Inc. Supplier No. R0619L
A restriction endonuclease that recognizes the sequence A^CG_T.