Learn More
New England Biolabs, Inc. HpyCH4V – 100 units
SDP
Catalog No. 50812360
Encompass
A restriction endonuclease that recognizes the sequence TG_^CA.
Catalog No. 50-812-360 Supplier New England Biolabs, Inc. Supplier No. R0620S
A restriction endonuclease that recognizes the sequence TG_^CA.