Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HQ-415 | 430462-93-4 | 98.1% | 415.48 | 1 ML
SDP

Catalog No. 5000388132
Encompass_Preferred

HQ-415 is a biologically active metal chelator primarily for research use. It is related to Clioquinol and exhibits an EC50 of 15 μM.

  • Biologically active metal chelator
  • Related to Clioquinol
  • EC50 of 15 μM
  • White to off-white solid appearance
  • Purity of 98.05%
  • Molecular weight of 415.48
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 2 years, -20°C for 1 year

Catalog No. 50-003-88132 Supplier Medchemexpress LLC Supplier No. HY1867010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HQ-415 is a biologically active metal chelator primarily for research use. It is related to Clioquinol and exhibits an EC50 of 15 μM.

  • Biologically active metal chelator
  • Related to Clioquinol
  • EC50 of 15 μM
  • White to off-white solid appearance
  • Purity of 98.05%
  • Molecular weight of 415.48
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.