Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HQ-415 | 430462-93-4 | 98.1% | 415.48 | C25H25N3O3 | 10 MG
SDP

Catalog No. 5000294910
Encompass_Preferred

HQ-415 is a biologically active metal chelator related to clioquinol, reported to have an EC50 of 15 μM. It is supplied as a research reagent for laboratory studies of metal chelation and related biochemical assays.

  • Biologically active metal chelator related to clioquinol.
  • Reported EC50 15 μM.
  • Molecular formula C25H25N3O3; molecular weight 415.48.
  • Purity 98.1%.
  • Available in small research pack sizes: 1 mg, 5 mg, 10 mg.
  • Storage: powder at -20°C for long-term; in solvent store at -80°C when possible.
  • For research use only; not for human or animal therapeutic use.

Catalog No. 50-002-94910 Supplier Medchemexpress LLC Supplier No. HY1867010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HQ-415 is a biologically active metal chelator related to clioquinol, reported to have an EC50 of 15 μM. It is supplied as a research reagent for laboratory studies of metal chelation and related biochemical assays.

  • Biologically active metal chelator related to clioquinol.
  • Reported EC50 15 μM.
  • Molecular formula C25H25N3O3; molecular weight 415.48.
  • Purity 98.1%.
  • Available in small research pack sizes: 1 mg, 5 mg, 10 mg.
  • Storage: powder at -20°C for long-term; in solvent store at -80°C when possible.
  • For research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.