Learn More
Hach Company HQ440D BOD and pH Meter Pkg
Catalog No. 5000207860 Shop All Hach Company Products
Encompass
HQD HQ440D Water Quality Laboratory Biochemical Oxygen Demand (BOD) and pH Meter Package-LBOD101 Sensor and PHC301 ElectrodeMeter package includes HQ440D Laboratory Multi Meter-Universal electrode Stand with electrode holder-PHC301 Standard Refillable pH Electrode with 1 m cable-pH Buffer Kit 4 01/7 00/10 01 (500 mL)-pH electrode storage solution (500 mL)-LBOD101 Luminescent/Optical Dissolved Oxygen (LDO) electrode for BOD Measurement with 1 m cable-Disposable 300 mL BOD Bottle (100 pcs)-BOD Bottle Stoppers (25 pcs)-BOD Bottle cap (6 pcs)-4 Alkaline 1 5V AA batteries-Power Supply Adapter-Power Cord-USB cable for data transfer-and Operating Documentation
Catalog No. 50-002-07860 Supplier Hach Company Supplier No. 8510100
Add to Cart
Add to Cart
Request a Quote
HQD HQ440D Water Quality Laboratory Biochemical Oxygen Demand (BOD) and pH Meter Package-LBOD101 Sensor and PHC301 ElectrodeMeter package includes HQ440D Laboratory Multi Meter-Universal electrode Stand with electrode holder-PHC301 Standard Refillable pH Electrode with 1 m cable-pH Buffer Kit 4 01/7 00/10 01 (500 mL)-pH electrode storage solution (500 mL)-LBOD101 Luminescent/Optical Dissolved Oxygen (LDO) electrode for BOD Measurement with 1 m cable-Disposable 300 mL BOD Bottle (100 pcs)-BOD Bottle Stoppers (25 pcs)-BOD Bottle cap (6 pcs)-4 Alkaline 1 5V AA batteries-Power Supply Adapter-Power Cord-USB cable for data transfer-and Operating Documentation