Learn More
Hach Company HQ440D pH and Cond. Meter Pkg
Supplier:  Hach Company 8507570
Encompass
HQD HQ440D General Purposes Water Laboratory pH and Conductivity/TDS Meter PackageMeter package includes HQ440D Laboratory Multi Meter-Universal electrode Stand with electrode holder-PHC201 combined pH gel electrode with 1 m cable-CDC401 conductivity/TDS/salinity cell with 1 m cable-pH Buffer Kit 4 01/7 00/10 01 (500 mL)-pH electrode storage solution (500 mL)-NaCl conductivity standard (1000 S/cm-491 mg/L-100 mL)-4 Alkaline 1 5V AA batteries-Power Supply Adapter-Power Cord-USB cable for data transfer-and Operating Documentation
Catalog No. 50-002-07841