Learn More
Medchemexpress LLC HY-108259 5mg Medchemexpress, HQL-79 CAS:162641-16-9 Purity:>98%
SDP
Catalog No. 501932314
Encompass_Preferred
Medchemexpress, HY-108259 5mg HQL-79 CAS:162641-16-9 HQL-79 is a potent, selective and orally active human hematopoietic prostaglandin D synthase (H-PGDS) inhibitor, highly selectively inhibits the synthesis of PGD 2 , and acts as an anti-allergic agent, with a K d of 0.8 μM and an IC 50 of 6 μM. Shows no obvious effect on COX-1, COX-2, m-PGES, or L-PGDS [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2314 Supplier Medchemexpress LLC Supplier No. HY1082595MG
Medchemexpress, HY-108259 5mg HQL-79 CAS:162641-16-9 HQL-79 is a potent, selective and orally active human hematopoietic prostaglandin D synthase (H-PGDS) inhibitor, highly selectively inhibits the synthesis of PGD 2 , and acts as an anti-allergic agent, with a K d of 0.8 μM and an IC 50 of 6 μM. Shows no obvious effect on COX-1, COX-2, m-PGES, or L-PGDS [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.