Learn More
TARGETMOL CHEMICALS INC HQNO 25MG
SDP
Catalog No. 502579073
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HQNO is a natural product isolated from P. aeruginosaand is a potent inhibitor of electron transport chain(Kd of 64 nM for complex III). It also is a potent mitochondrial NDH-2 inhibitorin many species. purity: 99%
Catalog No. 50-257-9073 Supplier TARGETMOL CHEMICALS INC Supplier No. T843325MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HQNO is a natural product isolated from P. aeruginosaand is a potent inhibitor of electron transport chain(Kd of 64 nM for complex III). It also is a potent mitochondrial NDH-2 inhibitorin many species. purity: 99%