Learn More
Aviva Systems Biology Corp HR Antibody - middle region (ARP32907_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32907_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HR. Alternate names: ALUNC, AU, HSA277165, MUHH, MUHH1ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-54814